missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium tungsten oxide, 99% min
Description
Lithium tungsten oxide is employed for the preparation of ceramics with ultra-low sintering temperatures. It acts as a catalyst for oxidative coupling reactions. Furthermore, it is used in the production of high density aqueous polytungstate (metatungstate) solutions which are utilized for the separation of minerals and other solids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 742°C |
| Physical Form | Powder |
| Quantity | 50 g |
| Solubility Information | Soluble in water. |
| Formula Weight | 261.73 |
| Percent Purity | ≥99% |
| Density | 3.71 g/mL |
| Chemical Name or Material | Lithium tungsten oxide |
Safety and Handling
| EINECSNumber | 236-978-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only