CAS RN 13568-45-1
CAS RN 13568-45-1
IUPAC Name:
dilithium(1+) dioxotungstenbis(olate)
Synonyms:
lithium tungstate
Molecular Weight (g/mol):
261.72
Molecular Formula:
Li2O4W
InChi Key:
SCKKBXOJPMZVPF-UHFFFAOYSA-N
SMILES:
[Li+].[Li+].[O-][W]([O-])(=O)=O
1
–
1
of
1
results
1
Lithium tungsten oxide, 99% min
CAS: 13568-45-1 Molecular Formula: Li2O4W Molecular Weight (g/mol): 261.716 MDL Number: MFCD00016185 InChI Key: SCKKBXOJPMZVPF-UHFFFAOYSA-N Synonym: lithium tungstate,dilithium dioxido dioxo tungsten,dilithotab tungstate,2li.wo4,dilithium 1+ ion tungstate PubChem CID: 6095624 IUPAC Name: dilithium;dioxido(dioxo)tungsten SMILES: [Li+].[Li+].[O-][W](=O)(=O)[O-]