missing translation for 'onlineSavingsMsg'
Learn More

Lithium (trimethylsilyl)acetylide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog No. p-3527070
Change view
Click to view available options
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC380658000 800 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC380658000 Supplier Thermo Scientific Chemicals Supplier No. 380658000
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54655-07-1 , 109-99-9
Molecular Formula C5H9LiSi
Molecular Weight (g/mol) 104.15
MDL Number MFCD00075059
InChI Key WDDOQHLJFOUQMW-UHFFFAOYSA-N
Synonym trimethylsilyl ethynyllithium, lithium, trimethylsilyl ethynyl, lithium 1+ ion trimethylsilyl ethyne, lithium trimethylsilylacetylide, lithio trimethylsilyl acetylene, lithium trimethylsilyl acetylide, lithium ethynyl trimethyl silane, lithium trimethylsilylacetylenide, zvxxeonxfwsciz-uhfffaoysa-n
PubChem CID 3431600
IUPAC Name lithium;ethynyl(trimethyl)silane
SMILES [Li+].C[Si](C)(C)C#[C-]

Specifications

Color Yellow to Orange
Physical Form Solution
Quantity 800 mL
Solubility Information Solubility in water: reacts
Formula Weight 104.15
Concentration or Composition (by Analyte or Components) 0.4 to 0.6M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.88
Density 0.8800g/mL
Chemical Name or Material Lithium (trimethylsilyl)acetylide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
May form explosive peroxides.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g

RUO – Research Use Only