missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Tetrakis(pentafluorophenyl)borate - Ethyl Ether Complex 70.0+%, TCI America™
$464.14 - $1388.71
Chemical Identifiers
| CAS | 155543-02-5 |
|---|---|
| Molecular Formula | C28H10BF20LiO |
| Molecular Weight (g/mol) | 760.105 |
| MDL Number | MFCD01861289 |
| InChI Key | KPLZKJQZPFREPG-UHFFFAOYSA-N |
| PubChem CID | 23701484 |
| IUPAC Name | lithium;ethoxyethane;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide |
| SMILES | [Li+].[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.CCOCC |
Chemical Identifiers
| 155543-02-5 | |
| 760.105 | |
| KPLZKJQZPFREPG-UHFFFAOYSA-N | |
| lithium;ethoxyethane;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide |
| C28H10BF20LiO | |
| MFCD01861289 | |
| 23701484 | |
| [Li+].[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.CCOCC |
Specifications
| 155543-02-5 | |
| MFCD01861289 | |
| 3175 | |
| [Li+].[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.CCOCC | |
| 760.105 | |
| ≥70.0% (T) | |
| Lithium Tetrakis(pentafluorophenyl)borate - Ethyl Ether Complex |
| C28H10BF20LiO | |
| 1 g | |
| KPLZKJQZPFREPG-UHFFFAOYSA-N | |
| lithium;ethoxyethane;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide | |
| 23701484 | |
| Crystalline Powder |
Safety and Handling
TSCA : No