missing translation for 'onlineSavingsMsg'
Learn More

Lithium tantalum oxide, 99.9% (metals basis)

CAS: 12031-66-2 | LiO3Ta | 235.88 g/mol

Supplier:  Thermo Scientific Chemicals 04183418

 View more versions of this product

Catalog No. AA4183418

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lithium tantalum oxide
Powder
(metals basis)
MFCD00016174
JNQQEOHHHGGZCY-UHFFFAOYSA-N
lithium;oxido(dioxo)tantalum
5148101
99.9%
12031-66-2
50 g
LiO3Ta
lithium tantalate, lithium oxido dioxo tantalum, lithium 1+ tantalumoylolate
[Li+].[O--].[O--].[O--].[Ta+5]
235.88
235.89
Odorless

Chemical Identifiers

12031-66-2
235.88
JNQQEOHHHGGZCY-UHFFFAOYSA-N
5148101
[Li+].[O--].[O--].[O--].[Ta+5]
LiO3Ta
MFCD00016174
lithium tantalate, lithium oxido dioxo tantalum, lithium 1+ tantalumoylolate
lithium;oxido(dioxo)tantalum
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 234-757-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only