missing translation for 'onlineSavingsMsg'
Learn More

Lithium tantalum oxide, 99.9% (metals basis)

CAS: 12031-66-2 | LiO3Ta | 235.88 g/mol

$699.05 - $699.05

Chemical Identifiers

CAS 12031-66-2
Molecular Formula LiO3Ta
Molecular Weight (g/mol) 235.88
MDL Number MFCD00016174
InChI Key JNQQEOHHHGGZCY-UHFFFAOYSA-N
Synonym lithium tantalate, lithium oxido dioxo tantalum, lithium 1+ tantalumoylolate
PubChem CID 5148101
IUPAC Name lithium;oxido(dioxo)tantalum
SMILES [Li+].[O--].[O--].[O--].[Ta+5]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4183418
View Documents
Thermo Scientific Chemicals
04183418
50 g
Each for $699.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12031-66-2
235.88
JNQQEOHHHGGZCY-UHFFFAOYSA-N
5148101
[Li+].[O--].[O--].[O--].[Ta+5]
LiO3Ta
MFCD00016174
lithium tantalate, lithium oxido dioxo tantalum, lithium 1+ tantalumoylolate
lithium;oxido(dioxo)tantalum

Specifications

12031-66-2
50 g
MFCD00016174
JNQQEOHHHGGZCY-UHFFFAOYSA-N
lithium;oxido(dioxo)tantalum
5148101
99.9%
Lithium tantalum oxide
Powder
LiO3Ta
lithium tantalate, lithium oxido dioxo tantalum, lithium 1+ tantalumoylolate
[Li+].[O--].[O--].[O--].[Ta+5]
235.88
235.89
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 234-757-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only