missing translation for 'onlineSavingsMsg'
Learn More

Lithium sulfate hydrate, Puratronic™, 99.996% (metals basis)

Catalog No. p-7049129
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1298306 5 g
AA1298314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1298306 Supplier Thermo Scientific Chemicals Supplier No. 01298306
Only null left

CAS: 10377-48-7 | Li2O4S | 109.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10377-48-7
Molecular Formula Li2O4S
Molecular Weight (g/mol) 109.94
MDL Number MFCD00011086
InChI Key INHCSSUBVCNVSK-UHFFFAOYSA-L
Synonym lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
PubChem CID 66320
ChEBI CHEBI:53474
SMILES [Li+].[Li+].[O-]S([O-])(=O)=O

Specifications

Melting Point 130°C (in -H2O)
Color White
Physical Form Crystalline
Quantity 5 g
Linear Formula Li2SO4·xH2O
Merck Index 14,5541
IUPAC Name dilithium;sulfate
Formula Weight 109.94 (Anhydrous)
Percent Purity 99.996%
Grade Puratronic
Density 2.06 g/mL
Assay (metals basis)
Chemical Name or Material Lithium sulfate hydrate
Show More Show Less
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 233-820-4
RTECSNumber OJ6419000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only