missing translation for 'onlineSavingsMsg'
Learn More

Lithium sulfate, anhydrous, 99.7% (metals basis)

Catalog No. p-4407588
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1340430 250 g
AA13404A1 1 kg
AA13404A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1340430 Supplier Thermo Scientific Chemicals Supplier No. 01340430
Only null left

CAS: 10377-48-7 | Li2O4S | 109.94 g/mol

Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 10377-48-7
Molecular Formula Li2O4S
Molecular Weight (g/mol) 109.94
MDL Number MFCD00011086
InChI Key INHCSSUBVCNVSK-UHFFFAOYSA-L
Synonym lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
PubChem CID 66320
ChEBI CHEBI:53474
SMILES [Li+].[Li+].[O-]S([O-])(=O)=O

Specifications

Melting Point 845°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.7% (Metal basis)
Solubility Information Soluble in water. Insoluble in acetone,ethanol and pyridine.
IUPAC Name dilithium(1+) sulfate
Formula Weight 109.94
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Density 2.22 g/cm3 at 20°C
Chemical Name or Material Lithium sulfate, anhydrous
Show More Show Less
Hazard Category H302-H319
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 233-820-4
RTECSNumber OJ6419000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only