missing translation for 'onlineSavingsMsg'
Learn More

Lithium sulfate, for analysis, anhydrous

Catalog No. AC447550010
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC447550010 1 kg
AC447552500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447550010 Supplier Thermo Scientific Chemicals Supplier No. 447550010
Only null left

CAS: 10377-48-7 | Li2O4S | 109.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10377-48-7
Molecular Formula Li2O4S
Molecular Weight (g/mol) 109.94
MDL Number MFCD00011086
InChI Key INHCSSUBVCNVSK-UHFFFAOYSA-L
Synonym lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
PubChem CID 66320
ChEBI CHEBI:53474
IUPAC Name dilithium;sulfate
SMILES [Li+].[Li+].[O-]S([O-])(=O)=O

Specifications

Melting Point 859.0°C
Quantity 1 kg
Linear Formula Li2SO4
Merck Index 15,5597
Solubility Information (10% in water) Clear colorless to slightly hazy
Formula Weight 109.94
Grade Analytical
Specific Gravity 2.22
Density 2.2200g/mL
Chemical Name or Material Lithium sulfate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr
EINECSNumber 233-820-4

RUO – Research Use Only