missing translation for 'onlineSavingsMsg'
Learn More

Lithium stearate

Catalog No. p-7050721
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
250 g
Catalog No. Quantity
AA3933530 250 g
AA39335A1 1 kg
2 options

Catalog No. AA3933530

Supplier: Thermo Scientific Chemicals 03933530

Only null left

CAS: 4485-12-5 | C9H11ClN3 | 196.66 g/mol

Lithium stearate is used as a thickener and gelling agent to prepare oils into lubricating greases. It is the active component of lithium grease. Further, it is used as lubricants, lubricant additives, plastic and rubber products. In addition to this, it is used as a stabilizing agent in the plastics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4485-12-5
MDL Number MFCD00042032

Specifications

Melting Point 220°C
Molecular Formula C9H11ClN3
Quantity 250 g
Synonym 2-1h-benzo d imidazol-2-yl ethanamine dihydrochloride, 2-1h-benzimidazol-2-yl ethanamine dihydrochloride, 1h-benzimidazole-2-ethanamine dihydrochloride, 2-2-aminoethyl benzoimidazole dihydrochloride, 2-1h-benzoimidazol-2-yl ethylamine dihydrochloride, 1h-benzimidazole-2-ethanamine, dihydrochloride, 1h-benzimidazole-ethanamine dihydrochloride, 2-2-aminoethyl benzoimidazoledihydrochloride, 2-1h-1,3-benzodiazol-2-yl ethanamine dihydrochloride, 2-benzimidazol-2-ylethylamine, chloride, chloride
Solubility Information Insoluble in water.
InChI Key VLIBSZCLVYXJTG-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)NC(=N2)CCN.Cl.Cl
IUPAC Name 2-(1H-benzimidazol-2-yl)ethanamine;dihydrochloride
Molecular Weight (g/mol) 196.66
PubChem CID 2788071
Formula Weight 290.41
Physical Form Powder
Chemical Name or Material Lithium stearate
Show More Show Less
Hazard Category H500
EINECSNumber 224-772-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only