accessibility menu, dialog, popup

UserName

LiH3COO

CAS RN 4485-12-5

IUPAC Name: lithium(1+) octadecanoate
Synonyms: lithium stearate
Molecular Weight (g/mol): 290.42
Molecular Formula: C18H35LiO2
InChi Key: HGPXWXLYXNVULB-UHFFFAOYSA-M
SMILES: [Li+].CCCCCCCCCCCCCCCCCC([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)