missing translation for 'onlineSavingsMsg'
Learn More

Lithium stearate

Catalog No. AA3933530
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3933530 250 g
AA39335A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3933530 Supplier Thermo Scientific Chemicals Supplier No. 03933530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4485-12-5 | C9H11ClN3 | 196.66 g/mol

Lithium stearate is used as a thickener and gelling agent to prepare oils into lubricating greases. It is the active component of lithium grease. Further, it is used as lubricants, lubricant additives, plastic and rubber products. In addition to this, it is used as a stabilizing agent in the plastics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4485-12-5
MDL Number MFCD00042032

Specifications

Melting Point 220°C
Molecular Formula C9H11ClN3
Quantity 250 g
Synonym 2-1h-benzo d imidazol-2-yl ethanamine dihydrochloride, 2-1h-benzimidazol-2-yl ethanamine dihydrochloride, 1h-benzimidazole-2-ethanamine dihydrochloride, 2-2-aminoethyl benzoimidazole dihydrochloride, 2-1h-benzoimidazol-2-yl ethylamine dihydrochloride, 1h-benzimidazole-2-ethanamine, dihydrochloride, 1h-benzimidazole-ethanamine dihydrochloride, 2-2-aminoethyl benzoimidazoledihydrochloride, 2-1h-1,3-benzodiazol-2-yl ethanamine dihydrochloride, 2-benzimidazol-2-ylethylamine, chloride, chloride
Solubility Information Insoluble in water.
InChI Key VLIBSZCLVYXJTG-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)NC(=N2)CCN.Cl.Cl
IUPAC Name 2-(1H-benzimidazol-2-yl)ethanamine;dihydrochloride
Molecular Weight (g/mol) 196.66
PubChem CID 2788071
Formula Weight 290.41
Physical Form Powder
Chemical Name or Material Lithium stearate
Show More Show Less
Hazard Category H500
EINECSNumber 224-772-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only