missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Stearate 90.0+%, TCI America™
$50.05 - $175.20
Chemical Identifiers
| CAS | 4485-12-5 |
|---|---|
| Molecular Formula | C18H35LiO2 |
| Molecular Weight (g/mol) | 290.42 |
| MDL Number | MFCD00042032 |
| InChI Key | HGPXWXLYXNVULB-UHFFFAOYSA-M |
| Synonym | lithium stearate, lithium octadecanoate, octadecanoic acid, lithium salt, stearic acid, lithium salt, lithalure, litholite, stavinor, lithium stearate, pure, stearic acid lithium salt, unii-p31mc94p70 |
| PubChem CID | 517357 |
| IUPAC Name | lithium;octadecanoate |
| SMILES | [Li+].CCCCCCCCCCCCCCCCCC(=O)[O-] |
Chemical Identifiers
| 4485-12-5 | |
| 290.42 | |
| HGPXWXLYXNVULB-UHFFFAOYSA-M | |
| 517357 | |
| [Li+].CCCCCCCCCCCCCCCCCC(=O)[O-] |
| C18H35LiO2 | |
| MFCD00042032 | |
| lithium stearate, lithium octadecanoate, octadecanoic acid, lithium salt, stearic acid, lithium salt, lithalure, litholite, stavinor, lithium stearate, pure, stearic acid lithium salt, unii-p31mc94p70 | |
| lithium;octadecanoate |
Specifications
| 4485-12-5 | |
| White | |
| MFCD00042032 | |
| lithium stearate, lithium octadecanoate, octadecanoic acid, lithium salt, stearic acid, lithium salt, lithalure, litholite, stavinor, lithium stearate, pure, stearic acid lithium salt, unii-p31mc94p70 | |
| [Li+].CCCCCCCCCCCCCCCCCC(=O)[O-] | |
| 290.42 | |
| 290.42 | |
| Crystalline Powder |
| 220°C | |
| C18H35LiO2 | |
| 25 g | |
| HGPXWXLYXNVULB-UHFFFAOYSA-M | |
| lithium;octadecanoate | |
| 517357 | |
| ≥90.0% (T) | |
| Lithium Stearate |
Safety and Handling
EINECSNumber : (2)-0611
RTECSNumber : WI4370000
TSCA : Yes