missing translation for 'onlineSavingsMsg'
Learn More

Lithium Stearate 90.0+%, TCI America™

Catalog No. S023725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S023725G 25 g
S0237500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S023725G Supplier TCI America Supplier No. S023725G

Chemical Identifiers

CAS 4485-12-5
Molecular Formula C18H35LiO2
Molecular Weight (g/mol) 290.42
MDL Number MFCD00042032
InChI Key HGPXWXLYXNVULB-UHFFFAOYSA-M
Synonym lithium stearate, lithium octadecanoate, octadecanoic acid, lithium salt, stearic acid, lithium salt, lithalure, litholite, stavinor, lithium stearate, pure, stearic acid lithium salt, unii-p31mc94p70
PubChem CID 517357
IUPAC Name lithium;octadecanoate
SMILES [Li+].CCCCCCCCCCCCCCCCCC(=O)[O-]

Specifications

Melting Point 220°C
Color White
Quantity 25 g
Formula Weight 290.42
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lithium Stearate
EINECSNumber (2)-0611
RTECSNumber WI4370000
TSCA Yes