missing translation for 'onlineSavingsMsg'
Learn More

Lithium stearate, 95%

CAS: 4485-12-5 | C18H35LiO2 | 290.41 g/mol

$55.87 - $167.85

Chemical Identifiers

CAS 4485-12-5
Molecular Formula C18H35LiO2
Molecular Weight (g/mol) 290.41
MDL Number MFCD00042032
InChI Key HGPXWXLYXNVULB-UHFFFAOYSA-M
Synonym lithium stearate, lithium octadecanoate, octadecanoic acid, lithium salt, stearic acid, lithium salt, lithalure, litholite, stavinor, lithium stearate, pure, stearic acid lithium salt, unii-p31mc94p70
PubChem CID 517357
IUPAC Name lithium;octadecanoate
SMILES [Li+].CCCCCCCCCCCCCCCCCC(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC413340250
SDS View Documents
Thermo Scientific Chemicals
413340250
25 g Glass bottle
Each for $55.87
Only null left
 
AC413341000
SDS View Documents
Thermo Scientific Chemicals
413341000
100 g Glass bottle
Each for $167.85
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4485-12-5
290.41
HGPXWXLYXNVULB-UHFFFAOYSA-M
517357
[Li+].CCCCCCCCCCCCCCCCCC(=O)[O-]
C18H35LiO2
MFCD00042032
lithium stearate, lithium octadecanoate, octadecanoic acid, lithium salt, stearic acid, lithium salt, lithalure, litholite, stavinor, lithium stearate, pure, stearic acid lithium salt, unii-p31mc94p70
lithium;octadecanoate

Specifications

4485-12-5
White
95%
C18H35LiO2
MFCD00042032
lithium stearate, lithium octadecanoate, octadecanoic acid, lithium salt, stearic acid, lithium salt, lithalure, litholite, stavinor, lithium stearate, pure, stearic acid lithium salt, unii-p31mc94p70
[Li+].CCCCCCCCCCCCCCCCCC(=O)[O-]
290.41
290.41
Powder
220°C
Authentic
Glass bottle
CH3(CH2)16COOLi
25 g
HGPXWXLYXNVULB-UHFFFAOYSA-M
lithium;octadecanoate
517357
95%
Lithium stearate, 95%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 224-772-5

RTECSNumber : WI4370000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only