missing translation for 'onlineSavingsMsg'
Learn More

Lithium stearate, 95%

Catalog No. AC413340250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413340250 25 g Glass bottle
AC413341000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413340250 Supplier Thermo Scientific Chemicals Supplier No. 413340250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4485-12-5 | C18H35LiO2 | 290.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4485-12-5
Molecular Formula C18H35LiO2
Molecular Weight (g/mol) 290.41
MDL Number MFCD00042032
InChI Key HGPXWXLYXNVULB-UHFFFAOYSA-M
Synonym lithium stearate, lithium octadecanoate, octadecanoic acid, lithium salt, stearic acid, lithium salt, lithalure, litholite, stavinor, lithium stearate, pure, stearic acid lithium salt, unii-p31mc94p70
PubChem CID 517357
IUPAC Name lithium;octadecanoate
SMILES [Li+].CCCCCCCCCCCCCCCCCC(=O)[O-]

Specifications

Melting Point 220°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH3(CH2)16COOLi
Quantity 25 g
Formula Weight 290.41
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Lithium stearate, 95%
Show More Show Less
EINECSNumber 224-772-5
RTECSNumber WI4370000
TSCA TSCA

RUO – Research Use Only