missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Nitrate, Honeywell
Reagent Grade
$647.35 - $647.35
Chemical Identifiers
| CAS | 7790-69-4 |
|---|---|
| Molecular Formula | LiNO3 |
| Molecular Weight (g/mol) | 68.94 |
| MDL Number | MFCD00011094 |
| InChI Key | IIPYXGDZVMZOAP-UHFFFAOYSA-N |
| Synonym | lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure |
| PubChem CID | 10129889 |
| ChEBI | CHEBI:63315 |
| IUPAC Name | lithium;nitrate |
| SMILES | [Li+].[O-][N+]([O-])=O |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
| 7790-69-4 | |
| 68.94 | |
| IIPYXGDZVMZOAP-UHFFFAOYSA-N | |
| 10129889 | |
| lithium;nitrate |
| LiNO3 | |
| MFCD00011094 | |
| lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure | |
| CHEBI:63315 | |
| [Li+].[O-][N+]([O-])=O |
Specifications
| 7790-69-4 | |
| LiNO3 | |
| MFCD00011094 | |
| lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure | |
| IIPYXGDZVMZOAP-UHFFFAOYSA-N | |
| lithium;nitrate | |
| 10129889 | |
| 68.95g/mol | |
| Lithium Nitrate |
| 1 kg | |
| LiNO3 | |
| 2722 | |
| Soluble in water | |
| [Li+].[O-][N+]([O-])=O | |
| 68.94 | |
| CHEBI:63315 | |
| ReagentPlus |
Safety and Handling
P210-P220-P221-P301 + P312 + P330-P305 + P351 + P338-P370 + P378
H272-H302-H319
EINECSNumber : 232-218-9
RTECSNumber : QU9330000