missing translation for 'onlineSavingsMsg'
Learn More

Lithium Nitrate, Solstice

Catalog No. 6000911
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000911 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000911 Supplier Solstice Supplier No. 2279861KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent Grade

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 7790-69-4
Molecular Formula LiNO3
Molecular Weight (g/mol) 68.94
MDL Number MFCD00011094
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
Synonym lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
PubChem CID 10129889
ChEBI CHEBI:63315
IUPAC Name lithium;nitrate
SMILES [Li+].[O-][N+]([O-])=O

Specifications

Quantity 1 kg
Linear Formula LiNO3
UN Number 2722
Solubility Information Soluble in water
Formula Weight 68.95g/mol
Grade ReagentPlus
Chemical Name or Material Lithium Nitrate
Hazard Statement P210-P220-P221-P301 + P312 + P330-P305 + P351 + P338-P370 + P378
Precautionary Statement H272-H302-H319
EINECSNumber 232-218-9
RTECSNumber QU9330000