missing translation for 'onlineSavingsMsg'
Learn More

Lithium isopropoxide, 3M solution in THF, AcroSeal™, Thermo Scientific™

Chemical Identifiers

CAS 2388-10-5 , 109-99-9
Molecular Formula CH3H7LiO
Molecular Weight (g/mol) 66.03
MDL Number MFCD00048286
InChI Key HAUKUGBTJXWQMF-UHFFFAOYSA-N
Synonym lithium isopropoxide, lithium propan-2-olate, lithium isopropanolate, lithium 2-propanolate, acmc-1ch0u, lithium isopropoxide solution, 2-propanol, lithiumsalt 1:1, lithium 1+ propan-2-olate
PubChem CID 23673350
IUPAC Name lithium;propan-2-olate
SMILES [Li+].CC(C)[O-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
missing translation for 'productCode' missing translation for 'brand' Quantity Packaging Price Quantity & Availability  
AC396508000
View Documents Promotion Details
N/A 800mL AcroSeal™ Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2388-10-5 , 109-99-9
66.03
HAUKUGBTJXWQMF-UHFFFAOYSA-N
23673350
[Li+].CC(C)[O-]
CH3H7LiO
MFCD00048286
lithium isopropoxide, lithium propan-2-olate, lithium isopropanolate, lithium 2-propanolate, acmc-1ch0u, lithium isopropoxide solution, 2-propanol, lithiumsalt 1:1, lithium 1+ propan-2-olate
lithium;propan-2-olate

Specifications

2388-10-5 , 109-99-9
LiOCH(CH3)2
−19°C
HAUKUGBTJXWQMF-UHFFFAOYSA-N
lithium;propan-2-olate
23673350
0.87
Lithium isopropoxide
CH3H7LiO
MFCD00048286
lithium isopropoxide, lithium propan-2-olate, lithium isopropanolate, lithium 2-propanolate, acmc-1ch0u, lithium isopropoxide solution, 2-propanol, lithiumsalt 1:1, lithium 1+ propan-2-olate
[Li+].CC(C)[O-]
66.03
66.03
0.8700g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Reacts violently with water.
May form explosive peroxides.
Suspected of causing cancer.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only