missing translation for 'onlineSavingsMsg'
Learn More

Lithium isobutyrate, 98+%

Catalog No. AA3937214
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3937214 25 g
AA3937236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3937214 Supplier Thermo Scientific Chemicals Supplier No. 03937214

CAS: 25179-23-1 | C4H7LiO2 | 94.038 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25179-23-1
Molecular Formula C4H7LiO2
Molecular Weight (g/mol) 94.038
MDL Number MFCD00210622
InChI Key IIDVGIFOWJJSIJ-UHFFFAOYSA-M
Synonym lithium isobutyrate, lithium 2-methylpropanoate, acmc-1ccd2, isobutyric acid lithium salt, lithium 1+ isobutyrate, lithium 1+ ion isobutyrate
PubChem CID 23665557
IUPAC Name lithium;2-methylpropanoate
SMILES [Li+].CC(C)C(=O)[O-]

Specifications

Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 94.03
Percent Purity ≥98%
Physical Form Powder or Lumps
Chemical Name or Material Lithium isobutyrate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only