missing translation for 'onlineSavingsMsg'
Learn More

Lithium Hexafluorophosphate 97.0+%, TCI America™

Catalog No. p-7131839
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L014625G 25 g
L0146100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L014625G Supplier TCI America Supplier No. L014625G
Only null left

Chemical Identifiers

CAS 21324-40-3
Molecular Formula F6LiP
Molecular Weight (g/mol) 151.904
MDL Number MFCD00011096
InChI Key AXPLOJNSKRXQPA-UHFFFAOYSA-N
Synonym lithium hexafluorophosphate, lithium hexafluorophosphate v, phosphate 1-, hexafluoro-, lithium, phosphate 1-, hexafluoro-, lithium 1:1, lithiumhexafluorophosphate, lithium hexafluorophosphate 1-, lipf6, lithium hexafluoroposphate, acmc-209fj7, ksc492g4h
PubChem CID 23688915
IUPAC Name lithium;hexafluorophosphate
SMILES [Li+].F[P-](F)(F)(F)(F)F

Specifications

Melting Point 200°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
UN Number 2923
Formula Weight 151.90
Percent Purity ≥97.0% (T)
Chemical Name or Material Lithium Hexafluorophosphate