Learn More
Lithium hexafluoroantimonate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03859514
Description
Lithium hexafluoroantimonate as a photo initiator for production of polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (decomposition) | |
| 18424-17-4 | |
| 25 g | |
| MFCD00042319 | |
| lithium hexafluoroantimonate, lithium 1+ ion hexafluorostibanuide, acmc-1bpt7, lithotab hexafluorostibanuide, lithium hexafluorostiboranuide, hexafluorostibine, lithium salt, lithium hexafluoroantimonate 1-, lithium hexakis fluoranyl antimony 1- | |
| YVBBFYDVCPCHHG-UHFFFAOYSA-H | |
| lithium;hexafluoroantimony(1-) | |
| 16700586 | |
| 97% |
| Lithium hexafluoroantimonate | |
| Powder | |
| F6LiSb | |
| UN1549 | |
| Very soluble in water | |
| [Li+].F[Sb-](F)(F)(F)(F)F | |
| 242.69 | |
| 242.68 | |
| Hygroscopic |
Chemical Identifiers
| 18424-17-4 | |
| 242.69 | |
| YVBBFYDVCPCHHG-UHFFFAOYSA-H | |
| 16700586 | |
| [Li+].F[Sb-](F)(F)(F)(F)F |
| F6LiSb | |
| MFCD00042319 | |
| lithium hexafluoroantimonate, lithium 1+ ion hexafluorostibanuide, acmc-1bpt7, lithotab hexafluorostibanuide, lithium hexafluorostiboranuide, hexafluorostibine, lithium salt, lithium hexafluoroantimonate 1-, lithium hexakis fluoranyl antimony 1- | |
| lithium;hexafluoroantimony(1-) |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only