accessibility menu, dialog, popup

UserName

CAS RN 18424-17-4

LiFSbFFFFF

CAS RN 18424-17-4

IUPAC Name: lithium(1+) hexafluorostibanuide
Synonyms: lithium(1+) hexafluorostibanuide
Molecular Weight (g/mol): 242.69
Molecular Formula: F6LiSb
InChi Key: YVBBFYDVCPCHHG-UHFFFAOYSA-H
SMILES: [Li+].F[Sb-](F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results