missing translation for 'onlineSavingsMsg'
Learn More

Lithium granules, 1-6mm (0.04-0.2in), 99%

Catalog No. AA0073318
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0073318 50 g
AA0073330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0073318 Supplier Thermo Scientific Chemicals Supplier No. 0073318

Sodium is major impurity | CAS: 7439-93-2 | Li | 6.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 7439-93-2
Molecular Formula Li
Molecular Weight (g/mol) 6.94
MDL Number MFCD00134051
InChI Key WHXSMMKQMYFTQS-UHFFFAOYSA-N
Synonym litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
PubChem CID 3028194
ChEBI CHEBI:30145
IUPAC Name lithium
SMILES [Li]

Specifications

Color Gray
Appearance Gray granules
Density 0.53g/cm3 at 20°C
Quantity 50 g
Odor Odorless
Packaging Under Argon
Form Granular
Assay Percent Range 99% (Metals basis)
Chemical Name or Material Lithium
Hazard Category H260-H314
Hazard Statement Keep container dry. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Precautionary Statement P223-P231+P232-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation DOT Class: 4.3, Packaging Group: I
EINECSNumber 231-102-5
RTECSNumber OJ5540000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only