missing translation for 'onlineSavingsMsg'
Learn More

Lithium DL-2-Hydroxybutyrate 98.0+%, TCI America™

Catalog No. H058025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H058025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H058025G Supplier TCI America Supplier No. H058025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 381716-41-2
Molecular Formula C4H7LiO3
Molecular Weight (g/mol) 110.037
MDL Number MFCD00040991
InChI Key CILZBARDXHWUSX-UHFFFAOYSA-M
Synonym DL-2-Hydroxybutyric Acid Lithium Salt
PubChem CID 23707041
IUPAC Name lithium;2-hydroxybutanoate
SMILES [Li+].CCC(C(=O)[O-])O

Specifications

Color White
Quantity 25 g
Formula Weight 110.04
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lithium DL-2-Hydroxybutyrate