accessibility menu, dialog, popup

UserName

LiH3COOOH

CAS RN 381716-41-2

IUPAC Name: lithium(1+) 2-hydroxybutanoate
Synonyms: lithium(1+) 2-hydroxybutyrate
Molecular Weight (g/mol): 110.04
Molecular Formula: C4H7LiO3
InChi Key: CILZBARDXHWUSX-UHFFFAOYNA-M
SMILES: [Li+].CCC(O)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results