missing translation for 'onlineSavingsMsg'
Learn More

Lithium diphenylphosphide, 0.5M solution in THF, AcroSeal™

Catalog No. AC433621000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433621000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC433621000 Supplier Thermo Scientific Chemicals Supplier No. 433621000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65567-06-8 | C12H10LiP | 192.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65567-06-8 , 109-99-9
Molecular Formula C12H10LiP
Molecular Weight (g/mol) 192.13
MDL Number MFCD00046054
InChI Key WKUYEGHEUWHKIU-UHFFFAOYSA-N
Synonym lithiodiphenylphosphine, lithium diphenylphosphide solution, diphenylphosphine lithium salt, lithium diphenylphosphanide, lipph2, lithiodiphenyl phosphide, lithium diphenylphosphine, diphenylphosphino lithium, diphenyl lithium phosphide
PubChem CID 3478053
IUPAC Name lithiodiphenylphosphane
SMILES [Li]P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Boiling Point 65.0°C to 67.0°C
Color Orange to Red
Physical Form Liquid
Quantity 100 mL
Flash Point −20°C
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.925
Density 0.9250g/mL
Formula Weight 192.12
Concentration or Composition (by Analyte or Components) 0.40 to 0.60M
Chemical Name or Material Lithium diphenylphosphide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Suspected of causing cancer.
May cause drowsiness or dizzines
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only