missing translation for 'onlineSavingsMsg'
Learn More

Lithium Citrate, Powder, 90%, Spectrum™ Chemical

$2271.29 - $2271.29

Chemical Identifiers

CAS 6080-58-6
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611314
View Documents
Spectrum Chemical Mfg Cor
L1113500GM
500 g Poly Bottle
Each for $2,271.29
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6080-58-6

Specifications

6080-58-6
8.0 to 9.0
90%
Poly Bottle
1
500 g
Reagent
White Powder or Fine Crystals
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.