missing translation for 'onlineSavingsMsg'
Learn More

Lithium Chloride, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical

Catalog No. 18603931
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
125 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603931 12 kg Poly Pail
18603925 25 g Amber Glass Bottle
18603926 125 g Amber Glass Bottle
18603928 500 g Amber Glass Bottle
18603927 6 x 125 g Amber Glass Bottle
18603930 2.5 kg Amber Glass Bottle
18603929 6 x 500 g Amber Glass Bottle
18603932 125 lb. Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18603931 Supplier Spectrum Chemical Mfg Cor Supplier No. L110912KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L1109, 7447-41-8, LiCl

Spectrum™ Chemical Lithium Chloride, Crystal, Reagent, ACS is a chemical compound that is extremely soluble in polar solvents and is used in order to obtain lithium metal and as a synthetic reagent. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Color White
CAS Max % 100%
Quantity 12 kg
Assay Percent Range 99%
Linear Formula LiCl
UN Number NONH for all modes of transport
Solubility Information Extremely soluble in polar solvents
InChI Formula InChI=1S/ClH.Li/h1H;/q;+1/p-1
PubChem CID 433294
Formula Weight 42.39
Percent Purity 99%
Grade Meets ACS reagent chemical requirements and Molecular Biology Grade specifications
Manufacturing Origin USA
Loss on Drying 1%
Odor Odorless
Packaging Poly Pail
Sensitivity Deliquescent
Appearance White crystals
Decomposition Information Hydrogen Chloride, Lithium Oxides
Chemical Name or Material Lithium Chloride
Show More Show Less