missing translation for 'onlineSavingsMsg'
Learn More

Lithium Chloride (Certified), Fisher Chemical™

Catalog No. p-32236
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
L121500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L121500 Supplier Thermo Fisher Scientific Supplier No. L121 500
Only null left

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Boiling Point 1382°C
Melting Point 605°C
Color White
pH 7 to 8
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥98.5 %
Formula Weight 42.39
Percent Purity ≥98.5%
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 1.33hPa at 547°C
Density 2.060g/cm³
Assay ≥98.5%
Chemical Name or Material Lithium Chloride
Show More Show Less