Learn More
Lithium chloride, anhydrous, 99%
CAS: 7447-41-8 | ClLi | 42.39 g/mol
$1954.19 - $1954.19
Chemical Identifiers
| CAS | 7447-41-8 |
|---|---|
| Molecular Formula | ClLi |
| Molecular Weight (g/mol) | 42.39 |
| MDL Number | MFCD00011078 |
| InChI Key | KWGKDLIKAYFUFQ-UHFFFAOYSA-M |
| Synonym | lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride |
| PubChem CID | 433294 |
| ChEBI | CHEBI:48607 |
| SMILES | [Li+].[Cl-] |
Description
Lithium chloride is useful for the production of lithium metal, and for the generation of Mn(0) species which can be used in free radical cyclizations. It can serve as a flame colorant to generate dark red flames, a brazing flux for aluminum in automobiles, a hygrometer, and a desiccant for drying air streams. On exposure to air, it becomes a solution with the concentration directly related to relative humidity of the atmosphere, and hence serves as a relative humidity standard in calibrating hygrometers. Apart from being a source of chloride, it serves as an additive in the Stille reaction in organic synthesis, and to precipitate RNA from cellular extracts. Being biologically significant, it finds applications in a wide variety of assays to study cell-fate and neurobiology. Lithium chloride has been found to inhibit virus infection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7447-41-8 | |
| 42.39 | |
| KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
| 433294 | |
| [Li+].[Cl-] |
| ClLi | |
| MFCD00011078 | |
| lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride | |
| CHEBI:48607 |
Specifications
| 1325°C to 1360°C | |
| 7447-41-8 | |
| Crystalline Powder | |
| 99% | |
| MFCD00011078 | |
| Freely soluble in water; soluble in methanol,ethanol,formic acid. Insoluble in dichloromethane. | |
| [Li+].[Cl-] | |
| 42.39 | |
| CHEBI:48607 | |
| Odorless | |
| 2.068 |
| 605°C | |
| White to Gray | |
| 5 kg | |
| ClLi | |
| lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride | |
| KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
| lithium;chloride | |
| 433294 | |
| 42.39 | |
| Hygroscopic | |
| Lithium chloride, anhydrous |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H302-H315-H319
EINECSNumber : 231-212-3
RTECSNumber : OJ5450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only