missing translation for 'onlineSavingsMsg'
Learn More

Lithium Chloride, ACS Reagent, Honeywell Fluka™

Catalog No. p-7071986
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002283 100 g
6002285 500 g
6002284 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6002283 Supplier Honeywell Chemicals Supplier No. 310468100G
Only null left

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium;chloride
SMILES [Li+].[Cl-]

Specifications

Quantity 100 g
Linear Formula LiCl
UN Number NONH for all modes of transport
Formula Weight 42.39g/mol
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Lithium Chloride
missing translation for 'healthHazard2' P301 + P312 + P330-P305 + P351 + P338
missing translation for 'healthHazard3' H302-H315-H319
EINECSNumber 231-212-3
RTECSNumber OJ5950000