missing translation for 'onlineSavingsMsg'
Learn More

Lithium chloride, ACS, 99% min

Catalog No. AA36217A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36217A3 2 kg
AA3621736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA36217A3 Supplier Thermo Scientific Chemicals Supplier No. 036217A3
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7447-41-8 | ClLi | 42.39 g/mol

In air-conditioning, welding and soldering flux, dry batteries, heat-exchange media, salt baths, and desiccants.Lithium chloride is useful for the production of lithium metal, and for the generation of Mn(0) species which can be used in free radical cyclizations. It can serve as a flame colorant to generate dark red flames, a brazing flux for aluminum in automobiles, a hygrometer, and a desiccant for drying air streams. On exposure to air, it becomes a solution with the concentration directly related to relative humidity of the atmosphere, and hence serves as a relative humidity standard in calibrating hygrometers. Apart from being a source of chloride, it serves as an additive in the Stille reaction in organic synthesis, and to precipitate RNA from cellular extracts. Being biologically significant, it finds applications in a wide variety of assays to study cell-fate and neurobiology. Lithium chloride has been found to inhibit virus infection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Boiling Point 1325°C to 1360°C
Melting Point 605°C
Physical Form Granules
Quantity 2 kg
Assay Percent Range 99%
Solubility Information Very soluble in water,alcohol,ether,pyridine,and nitrobenzene.
Formula Weight 42.39
Grade ACS Reagent
Sensitivity Hygroscopic
Density 2.068 g/mL
Chemical Name or Material Lithium Chloride
Show More Show Less
Hazard Category H302-H315-H319
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
EINECSNumber 231-212-3
RTECSNumber OJ5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only