missing translation for 'onlineSavingsMsg'
Learn More

Lithium Chloride, 99% (ACS Grade), MP Biomedicals

Catalog No. ICN15253890
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15253890 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15253890 Supplier MP Biomedicals Inc Supplier No. 0215253890

missing translation for 'validMainframeMsg'

ACS reagent grade lithium chloride for analytical applications with 99+% purity.

  • Grade: ACS reagent
  • Purity: 99+%

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99+%
Linear Formula LiCl
UN Number NONH for all modes of transport
Solubility Information Soluble in water
InChI Formula InChI=1S/ClH.Li/h1H;/q;+1/p-1
Formula Weight 42.4
Percent Purity ≥99%
Grade ACS
Manufacturing Origin USA
Identification Passes Test
Decomposition Information Hydrogen Chloride; Lithium Oxides
Chemical Name or Material Lithium Chloride, 99%
Show More Show Less