missing translation for 'onlineSavingsMsg'
Learn More

Lithium Chloride (2.3% in Tetrahydrofuran, ca. 0.5mol/L), TCI America™

Supplier:  TCI America L0222100ML

Catalog No. L0222100ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Lithium Chloride (2.3% in Tetrahydrofuran, ca. 0.5mol/L)
Yellow
MFCD00011078
2056
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium(1+) chloride
433294
42.39
7447-41-8
ClLi
100 mL
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
[Li+].[Cl-]
42.39
CHEBI:48607
Liquid

Chemical Identifiers

7447-41-8
42.39
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
433294
lithium(1+) chloride
ClLi
MFCD00011078
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
CHEBI:48607
[Li+].[Cl-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (1)-0231

RTECSNumber : OJ5950000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions