missing translation for 'onlineSavingsMsg'
Learn More

Lithium chloride, 1M aq. soln.

Catalog No. p-7042258
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61895AD 50 mL
AAJ61895AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61895AD Supplier Thermo Scientific Chemicals Supplier No. J61895AD
Only null left

CAS: 7447-41-8 | ClLi | 42.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
SMILES [Li+].[Cl-]

Specifications

Color Colorless
Physical Form Liquid
Quantity 50 mL
Linear Formula LiCl
IUPAC Name lithium;chloride
Formula Weight 42.39
Concentration or Composition (by Analyte or Components) 1M aq. soln.
Odor Odorless
Chemical Name or Material Lithium chloride
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only