missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Carbonate, Honeywell Fluka™
Purum,≥99%
Supplier: Honeywell Chemicals 13010100KG
Description
- ≥95% for general laboratory use.
Specifications
| Lithium Carbonate | |
| 100 kg | |
| Li2CO3 | |
| NONH for all modes of transport | |
| lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine | |
| [Li+].[Li+].[O-]C([O-])=O | |
| 73.89 | |
| CHEBI:6504 | |
| ≥99% |
| 554-13-2 | |
| CLi2O3 | |
| MFCD00011084 | |
| 3999191 | |
| XGZVUEUWXADBQD-UHFFFAOYSA-L | |
| dilithium;carbonate | |
| 11125 | |
| 73.89g/mol | |
| purum |
Chemical Identifiers
| 554-13-2 | |
| 73.89 | |
| XGZVUEUWXADBQD-UHFFFAOYSA-L | |
| 11125 | |
| dilithium;carbonate |
| CLi2O3 | |
| MFCD00011084 | |
| lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine | |
| CHEBI:6504 | |
| [Li+].[Li+].[O-]C([O-])=O |
Safety and Handling
P301 + P312 + P330-P305 + P351 + P338
H302-H319
EINECSNumber : 209-062-5
RTECSNumber : OJ5800000