missing translation for 'onlineSavingsMsg'
Learn More

Lithium Carbonate, Honeywell Fluka™

Catalog No. p-7071795
Change view
Click to view available options
Quantity:
1 kg
100 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001962 1 kg
6001961 100 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001962 Supplier Honeywell Chemicals Supplier No. 130101KG
Only null left

Purum,≥99%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium;carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Quantity 1 kg
Linear Formula Li2CO3
UN Number NONH for all modes of transport
Beilstein 3999191
Formula Weight 73.89g/mol
Percent Purity ≥99%
Grade purum
Chemical Name or Material Lithium Carbonate
Hazard Statement P301 + P312 + P330-P305 + P351 + P338
Precautionary Statement H302-H319
EINECSNumber 209-062-5
RTECSNumber OJ5800000