missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Lithium Buffer 5, pH 3.55

Catalog No. BC80203769
Change view
Click to view available options
Quantity:
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BC80203769 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BC80203769 Supplier Biochrom Supplier No. 80203769
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biochrom Amino Acid Analysis reagents and buffers are manufactured from high quality chemicals and materials and subjected to rigorous control procedures, under the ISO 13485 Quality System.

Biochrom buffers are guaranteed to give accurate and reproducible results for up to 3 years from manufacture, provided the storage conditions are adhered to. All reagents have stable chemistry and have been formulated to be stored at ambient temperature so, conveniently, do not need refrigeration.

Specifications

pH Range 3.55
Product Type Buffer
Content And Storage Store at ambient temperature
Quantity 2 L
Form Liquid