missing translation for 'onlineSavingsMsg'
Learn More

Lithium Bromide, 1.52 g/mL, Reagents

Catalog No. 01334828
Change view
Click to view available options
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01334828 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334828 Supplier Labchem Inc Supplier No. CL1965001A

Lithium Bromide, 1.52 g/mL, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7550-35-8 , 7732-18-5
Molecular Formula BrLi
Molecular Weight (g/mol) 86.84
InChI Key AMXOYNBUYSYVKV-UHFFFAOYSA-M
Synonym Lithium monobromide
IUPAC Name lithium(1+) bromide
SMILES [Li+].[Br-]

Specifications

Boiling Point 118°C
Color Colorless
Physical Form Liquid
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.5525
Chemical Name or Material Lithium Bromide, 1.52 g/mL