missing translation for 'onlineSavingsMsg'
Learn More

Lithium borohydride, 95%

Catalog No. p-44259
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC206810050 5 g
AC206810100 10 g
AC206810500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC206810050 Supplier Thermo Scientific Chemicals Supplier No. 206810050

CAS: 16949-15-8 | BH4Li | 21.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 16949-15-8
Molecular Formula BH4Li
Molecular Weight (g/mol) 21.78
MDL Number MFCD00011088
InChI Key UUKMSDRCXNLYOO-UHFFFAOYSA-N
Synonym borate 1-, tetrahydro-, lithium, unii-8l87x4s4kp, boryllithium, lithium, boryl, lithium hydroborate, lithotab borohydride, bh4li, lithium 1+ ion boranuide, lithium borohydride 2.0 m in thf, borate 1-, tetrahydro-, lithium 1:1
PubChem CID 20722760
SMILES [Li+].[BH4-]

Specifications

Melting Point 280.0°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 95%
Linear Formula LiBH4
Fieser 01,603; 04,296; 11,293; 12,276; 13,92; 14,191; 15,186
Merck Index 15, 5581
Solubility Information Solubility in water: 209g/L (10°C). Other solubilities: soluble in ether, ethanol, tetrahydrofuran and, aliphatic amines
IUPAC Name lithium(1+) boranuide
Formula Weight 21.77
Percent Purity 95%
Packaging Glass Bottle
Specific Gravity 0.66
Density 0.6600g/mL
Chemical Name or Material Lithium borohydride, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Reacts violentl
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face p
EINECSNumber 241-021-7
RTECSNumber ED2725000
TSCA TSCA

RUO – Research Use Only