missing translation for 'onlineSavingsMsg'
Learn More

Lithium Bis(trifluorométhansulfonyl)imide 98,0+%, TCI America™

Catalog No. B254225G
Change view
Click to view available options
Quantité:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
B254225G 25 g
B2542250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B254225G Supplier TCI America Supplier No. B254225G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90076-65-6
Formule moléculaire C2F6LiNO4S2
Poids moléculaire (g/mol) 287.075
Clé InChI QSZMZKBZAYQGRS-UHFFFAOYSA-N
Synonyme lithium bis trifluoromethanesulfonyl imide, lithium bis trifluoromethyl sulfonyl amide, lithium bis trifluoromethanesulphonyl imide, litfsi, lithium bis trifluoromethylsulfonyl azanide, lithium bis trifluoromethylsulfonyl imide, bis trifluoromethane sulfonimide lithium salt, bistrifluoromethanesulfonimide lithium salt, lithium bis trifluoromethanesulfonyl amide, lithium triflimide
PubChem CID 3816071
Nom de l’IUPAC lithium; bis(trifluorométhylsulfonyl)azanide
SOURIRES [Li+].C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Nom ou substance chimique Lithium Bis(trifluoromethanesulfonyl)imide
Point de fusion 235 °C
Couleur Blanc
Quantité 25 g
Numéro UN 2923
Poids de formule 287.08
Pourcentage de pureté ≥98,0% (T)
Forme physique Poudre cristalline
EINECSNumber (2)-3831
TSCA Oui