accessibility menu, dialog, popup

UserName

CAS RN 90076-65-6

IUPAC Name: lithium(1+) trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: lithium(1+) bistriflylimide anion
Molecular Weight (g/mol): 287.08
Molecular Formula: C2F6LiNO4S2
InChi Key: QSZMZKBZAYQGRS-UHFFFAOYSA-N
SMILES: [Li+].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (4)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)