missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Bis(pentafluoroethanesulfonyl)imide 98.0+%, TCI America™
$223.68 - $620.73
Chemical Identifiers
| CAS | 132843-44-8 |
|---|---|
| Molecular Formula | C4F10LiNO4S2 |
| Molecular Weight (g/mol) | 387.09 |
| MDL Number | MFCD22374096 |
| InChI Key | ACFSQHQYDZIPRL-UHFFFAOYSA-N |
| Synonym | Bis(pentafluoroethanesulfonyl)imide Lithium Salt |
| PubChem CID | 12096715 |
| IUPAC Name | lithium(1+) 1,1,1,2,2-pentafluoro-2-{[(1,1,2,2,2-pentafluoroethanesulfonyl)azanidyl]sulfonyl}ethane |
| SMILES | [Li+].FC(F)(F)C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)C(F)(F)F |
Chemical Identifiers
| 132843-44-8 | |
| 387.09 | |
| ACFSQHQYDZIPRL-UHFFFAOYSA-N | |
| 12096715 | |
| [Li+].FC(F)(F)C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)C(F)(F)F |
| C4F10LiNO4S2 | |
| MFCD22374096 | |
| Bis(pentafluoroethanesulfonyl)imide Lithium Salt | |
| lithium(1+) 1,1,1,2,2-pentafluoro-2-{[(1,1,2,2,2-pentafluoroethanesulfonyl)azanidyl]sulfonyl}ethane |
Specifications
| 132843-44-8 | |
| White | |
| MFCD22374096 | |
| 2923 | |
| ACFSQHQYDZIPRL-UHFFFAOYSA-N | |
| lithium(1+) 1,1,1,2,2-pentafluoro-2-{[(1,1,2,2,2-pentafluoroethanesulfonyl)azanidyl]sulfonyl}ethane | |
| 12096715 | |
| ≥98.0% (T) | |
| Lithium Bis(pentafluoroethanesulfonyl)imide |
| 328°C | |
| C4F10LiNO4S2 | |
| 1 g | |
| Bis(pentafluoroethanesulfonyl)imide Lithium Salt | |
| [Li+].FC(F)(F)C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)C(F)(F)F | |
| 387.09 | |
| 387.09 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-3839
TSCA : Yes