accessibility menu, dialog, popup

UserName

LiFFSOONSOOFFFFFFFF

CAS RN 132843-44-8

IUPAC Name: lithium(1+) 1,1,1,2,2-pentafluoro-2-{[(1,1,2,2,2-pentafluoroethanesulfonyl)azanidyl]sulfonyl}ethane
Synonyms: lithium(1+) 1,1,1,2,2-pentafluoro-2-[(1,1,2,2,2-pentafluoroethanesulfonyl)azanidylsulfonyl]ethane
Molecular Weight (g/mol): 387.09
Molecular Formula: C4F10LiNO4S2
InChi Key: ACFSQHQYDZIPRL-UHFFFAOYSA-N
SMILES: [Li+].FC(F)(F)C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results