missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Pig Lithium Battery Absorbent Mat Pad
Catch drips and leaks from lithium-ion cells using this chemical-resistant mat pad designed to safely absorb lithium-ion battery electrolyte
$61.28 - $227.99
Specifications
| Color | Pink |
|---|---|
| Material | Polypropylene |
| Type | Absorbent Mat |
| Disposable | Single-use |
| Weight Capacity | Lightweight |
| Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity | |||||
|
17-987-838
|
New Pig
MAT3102 |
50 Pk. |
N/A
|
|
|||||
|
17-987-839
|
New Pig
MAT3103 |
8 Pk. |
N/A
|
|
|||||
Description
- Chemical-Resistant Material:Specifically engineered to safely absorb the volatile and hazardous electrolytes found in lithium-ion batteries without degrading or causing a dangerous reaction.
- Durable Construction: Made from multiple layers of thermally bonded polypropylene, the pads are tear-resistant and won't rip, fray, or fall apart even when fully saturated, ensuring they hold up during use.
- Clear Identification: The pads are a distinct pink color, a visual cue that clearly marks them for use with hazardous chemicals and lithium-ion battery spills to prevent accidental misuse.
- Flame-Resistant: In its unused state, the mat is flame resistant and is designed to melt and self-extinguish when exposed to fire, adding a layer of safety during a thermal event.
- Convenient Pad Format: Unlike rolls, the pre-cut pad format is ideal for catching drips, soaking up small spills, and handling specific cleanup tasks without the need for cutting, which helps reduce waste.
Specifications
| Pink | |
| Absorbent Mat | |
| Lightweight |
| Polypropylene | |
| Single-use |