missing translation for 'onlineSavingsMsg'
Learn More

Lithium azide, pure, 20% solution in water

CAS: 19597-69-4 | LiN3

$231.31 - $643.51

Chemical Identifiers

CAS 19597-69-4
Molecular Formula LiN3
MDL Number MFCD00041956
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC335360250
SDS View Documents
Thermo Scientific Chemicals
335360250
25 g Glass bottle
Each for $231.31
Only null left
 
AC335361000
SDS View Documents
Thermo Scientific Chemicals
335361000
100 g Glass bottle
Each for $643.51
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, For Azidation/Diazo transfer
Specifications

Chemical Identifiers

19597-69-4
LiN3
MFCD00041956

Specifications

Colorless
LiN3
MFCD00041956
48.96
Pure
25 g
Lithium azide, 20% Solution in Water
19597-69-4
LiN3
1.0880g/mL
17 to 23%
Glass bottle
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Fatal if swallowed.
Contact with acids liberates very toxic gas.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 243-177-1

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only