Learn More
Lithium amide, 95%
CAS: 7782-89-0 | H2LiN | 22.963 g/mol
$69.73 - $503.11
Chemical Identifiers
| CAS | 7782-89-0 |
|---|---|
| Molecular Formula | H2LiN |
| Molecular Weight (g/mol) | 22.963 |
| MDL Number | MFCD00011093 |
| InChI Key | AFRJJFRNGGLMDW-UHFFFAOYSA-N |
| Synonym | lithium amide, lithamide, lithium amide li nh2, lithiumamide, unii-7393omu9lk, lithium azanide, hsdb 649, linh2, lithium amide 25g, lithium amide, powder |
| PubChem CID | 24532 |
| IUPAC Name | lithium;azanide |
| SMILES | [Li+].[NH2-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1602314
|
Thermo Scientific Chemicals
L1602314 |
25 g |
Each for $69.73
|
|
|||||
|
AAL1602322
|
Thermo Scientific Chemicals
L1602322 |
100 g |
Each for $133.64
|
|
|||||
|
AAL1602336
|
Thermo Scientific Chemicals
L1602336 |
500 g |
Each for $503.11
|
|
|||||
Description
Lithium amide is used in the preparation of active pharmaceutical ingredients and antioxidants. It acts as a catalyst for polymers, as nucleophiles and as strong bases. It serves as a reagent in the synthesis of antiinflamatory and preoresolving protectin D1, chemotype dipeptidyl peptidase IV inhibitors and sterically congested triarylamines. It finds application in dyes displaying large stokes shifts. In addition to this, it is used as a reagent for cross-coupling of aryl chlorides and amine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsLithium amide is used in the preparation of active pharmaceutical ingredients and antioxidants. It acts as a catalyst for polymers, as nucleophiles and as strong bases. It serves as a reagent in the synthesis of antiinflamatory and preoresolving protectin D1, chemotype dipeptidyl peptidase IV inhibitors and sterically congested triarylamines. It finds application in dyes displaying large stokes shifts. In addition to this, it is used as a reagent for cross-coupling of aryl chlorides and amine.
Solubility
Slightly soluble in ethanol and liquid ammonia. Insoluble in anhydrous ether, benzene and toluene.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids, water and alcohols.
Chemical Identifiers
| 7782-89-0 | |
| 22.963 | |
| AFRJJFRNGGLMDW-UHFFFAOYSA-N | |
| 24532 | |
| [Li+].[NH2-] |
| H2LiN | |
| MFCD00011093 | |
| lithium amide, lithamide, lithium amide li nh2, lithiumamide, unii-7393omu9lk, lithium azanide, hsdb 649, linh2, lithium amide 25g, lithium amide, powder | |
| lithium;azanide |
Specifications
| 430°C | |
| 7782-89-0 | |
| H2LiN | |
| MFCD00011093 | |
| 14,5522 | |
| Slightly soluble in ethanol and liquid ammonia. Insoluble in anhydrous ether,benzene and toluene. | |
| [Li+].[NH2-] | |
| 22.963 | |
| 22.96 | |
| Air and moisture sensitive | |
| Lithium amide |
| 373°C | |
| 25 g | |
| LiNH2 | |
| UN1390 | |
| lithium amide, lithamide, lithium amide li nh2, lithiumamide, unii-7393omu9lk, lithium azanide, hsdb 649, linh2, lithium amide 25g, lithium amide, powder | |
| AFRJJFRNGGLMDW-UHFFFAOYSA-N | |
| lithium;azanide | |
| 24532 | |
| 95% | |
| 1.178 |
Safety and Handling
GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P223-P231+P232-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H261-H314
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ALKALI METAL AMIDES
EINECSNumber : 231-968-4
RTECSNumber : OJ5590000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only