missing translation for 'onlineSavingsMsg'
Learn More

Lithium amide, 95%

CAS: 7782-89-0 | H2LiN | 22.963 g/mol

Supplier:  Thermo Scientific Chemicals L1602322

 View more versions of this product

Catalog No. AAL1602322

Only null left
Explore available promotions

Description

Description

Lithium amide is used in the preparation of active pharmaceutical ingredients and antioxidants. It acts as a catalyst for polymers, as nucleophiles and as strong bases. It serves as a reagent in the synthesis of antiinflamatory and preoresolving protectin D1, chemotype dipeptidyl peptidase IV inhibitors and sterically congested triarylamines. It finds application in dyes displaying large stokes shifts. In addition to this, it is used as a reagent for cross-coupling of aryl chlorides and amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium amide is used in the preparation of active pharmaceutical ingredients and antioxidants. It acts as a catalyst for polymers, as nucleophiles and as strong bases. It serves as a reagent in the synthesis of antiinflamatory and preoresolving protectin D1, chemotype dipeptidyl peptidase IV inhibitors and sterically congested triarylamines. It finds application in dyes displaying large stokes shifts. In addition to this, it is used as a reagent for cross-coupling of aryl chlorides and amine.

Solubility
Slightly soluble in ethanol and liquid ammonia. Insoluble in anhydrous ether, benzene and toluene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids, water and alcohols.
Specifications

Specifications

430°C
373°C
100 g
LiNH2
UN1390
lithium amide, lithamide, lithium amide li nh2, lithiumamide, unii-7393omu9lk, lithium azanide, hsdb 649, linh2, lithium amide 25g, lithium amide, powder
AFRJJFRNGGLMDW-UHFFFAOYSA-N
lithium;azanide
24532
95%
1.178
Lithium amide
7782-89-0
H2LiN
MFCD00011093
14,5522
Slightly soluble in ethanol and liquid ammonia. Insoluble in anhydrous ether,benzene and toluene.
[Li+].[NH2-]
22.963
22.96
Air and moisture sensitive

Chemical Identifiers

7782-89-0
22.963
AFRJJFRNGGLMDW-UHFFFAOYSA-N
24532
[Li+].[NH2-]
H2LiN
MFCD00011093
lithium amide, lithamide, lithium amide li nh2, lithiumamide, unii-7393omu9lk, lithium azanide, hsdb 649, linh2, lithium amide 25g, lithium amide, powder
lithium;azanide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P223-P231+P232-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H261-H314

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ALKALI METAL AMIDES

EINECSNumber : 231-968-4

RTECSNumber : OJ5590000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only