missing translation for 'onlineSavingsMsg'
Learn More

Lithium Aluminum Hydride (Powder) 95.0+%, TCI America™

Supplier:  TCI America L0203100G

 View more versions of this product

Catalog No. L0203100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

White
125°C
AlH4Li
1410
lithium aluminum hydride, lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
[Li+].[AlH4-]
37.95
37.95
Crystalline Powder
Lithium Aluminum Hydride (Powder)
16853-85-3
MFCD00011075
100 g
OCZDCIYGECBNKL-UHFFFAOYSA-N
lithium(1+) alumanuide
21226445
≥98.0% (T)

Chemical Identifiers

16853-85-3
37.95
OCZDCIYGECBNKL-UHFFFAOYSA-N
21226445
[Li+].[AlH4-]
AlH4Li
MFCD00011075
lithium aluminum hydride, lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
lithium(1+) alumanuide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (1)-0016

RTECSNumber : BD0100000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions